Assay Details
| Gene Symbol: Rps6ka4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019924.2 | NP_064308.2 | 6 - 7 | 734 | 74 | |
| GenBank mRNA | AF074714.1 | - | 6 - 7 | 706 | 74 | |
| AK089467.1 | - | 6 - 7 | 725 | 74 | ||
| AK154674.1 | - | 6 - 7 | 733 | 74 | ||
| BC012964.1 | - | 6 - 7 | 744 | 4160873 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka4
Genomic Map