Assay Details
| Gene Symbol: Rps6ka4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019924.2 | NP_064308.2 | 10 - 11 | 1280 | 79 | |
| GenBank mRNA | AF074714.1 | - | 10 - 11 | 1252 | 79 | |
| AK089467.1 | - | 10 - 11 | 1272 | 79 | ||
| AK154674.1 | - | 10 - 11 | 1279 | 79 | ||
| AK216854.1 | - | 1 - 2 | 37 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka4
Genomic Map