Assay Details
| Gene Symbol: Rps6ka4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019924.2 | NP_064308.2 | 11 - 12 | 1418 | 64 | |
| GenBank mRNA | AF074714.1 | - | 11 - 12 | 1390 | 64 | |
| AK089467.1 | - | 11 - 12 | 1410 | 64 | ||
| AK154674.1 | - | 11 - 12 | 1417 | 64 | ||
More Information
Set Membership: |
|

Rps6ka4
Genomic Map