Assay Details
| Gene Symbol: Mtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164190.1 | NP_001157662.1 | 7 - 8 | 719 | 109 | |
| NM_001164191.1 | NP_001157663.1 | 7 - 8 | 610 | 109 | ||
| NM_001164192.1 | NP_001157664.1 | 7 - 8 | 610 | 109 | ||
| NM_019926.3 | NP_064310.2 | 7 - 8 | 599 | 109 | ||
| GenBank mRNA | AF073996.1 | - | 7 - 8 | 600 | 109 | |
| AK016445.1 | - | 7 - 8 | 720 | 109 | ||
| AK031794.1 | - | 7 - 8 | 581 | 109 | ||
| AK166452.1 | - | 7 - 8 | 578 | 109 | ||
| AK171475.1 | - | 7 - 8 | 588 | 109 | ||
| BC051022.1 | - | 7 - 8 | 568 | 109 | ||
| BC066847.1 | - | 7 - 8 | 557 | 109 | ||
| BC090984.1 | - | 7 - 8 | 577 | 30246206 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtm1
Genomic Map