Assay Details
| Gene Symbol: Slc25a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020520.4 | NP_065266.1 | 5 - 6 | 664 | 104 | |
| GenBank mRNA | AK075624.1 | - | 5 - 6 | 665 | 104 | |
| AK152436.1 | - | 5 - 6 | 635 | 104 | ||
| BC029733.1 | - | 5 - 6 | 661 | 4019406 | 104 | |
| BC052871.1 | - | 5 - 6 | 587 | 30032324 | 104 | |
More Information
Set Membership: |
|

Slc25a20
Genomic Map