Assay Details
| Gene Symbol: Mrps31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020560.2 | NP_065585.1 | 5 - 6 | 907 | 92 | |
| GenBank mRNA | AK005110.1 | - | 5 - 6 | 866 | 92 | |
| AK011825.1 | - | 5 - 6 | 863 | 92 | ||
| AK161913.1 | - | 5 - 6 | 850 | 92 | ||
| BC006764.1 | - | 5 - 6 | 800 | 92 | ||
| BC027430.1 | - | 5 - 6 | 811 | 4187438 | 92 | |
| Z46966.1 | - | 5 - 6 | 826 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrps31
Genomic Map