Assay Details
| Gene Symbol: Snrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021336.4 | NP_067311.4 | 2 - 3 | 506 | 90 | |
| GenBank mRNA | AF230356.1 | - | 2 - 3 | 235 | 90 | |
| AK005256.1 | - | 2 - 3 | 298 | 90 | ||
| AK088438.1 | - | 2 - 3 | 281 | 90 | ||
| AK151800.1 | - | 2 - 3 | 299 | 90 | ||
| AK152546.1 | - | 2 - 3 | 299 | 90 | ||
| AK153502.1 | - | 2 - 3 | 299 | 90 | ||
| BC029642.1 | - | 2 - 3 | 273 | 3588138 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186097 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186101 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113208 | AK153502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113208 | BC013777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113208 | BC029642.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113208 | AK005256.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113208 | NM_021336.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120652 | NM_021336.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618843 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340796 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773673 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 911224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 179882 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156788 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151646 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113208 | AK153502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113208 | BC013777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113208 | BC029642.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113208 | AK005256.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113208 | NM_021336.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120652 | NM_021336.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25186104 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01534501 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151646 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156788 | - | - |
| Whole Mouse Genome G4122A | A_51_P164270 | AK153502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164270 | AK206837.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164270 | BC013777.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164270 | BC029642.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164270 | AF230356.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105703 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108969 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156788 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151646 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06332868 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06299199 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186097 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25186100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25186175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06401694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06460394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06309474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06201438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06460388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06018085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25186125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06271718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10553996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553998 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554001 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554000 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553997 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417353_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417352_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10553993 | - | - |
| Mouse Genome 430 2.0 Array | 1417351_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417352_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1417353_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1417352_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1417351_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417353_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68981_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68981_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700009099.mm.1 | NM_021336.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009099.mm.1 | NM_021336.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009099.mm.1 | TC0700000989.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417353_x_at | - | - |
| Mouse Expression Set 430 | 1417352_PM_s_at | - | - |
| Mouse Expression Set 430 | 1417352_s_at | - | - |
| Mouse Expression Set 430 | 1417353_PM_x_at | - | - |
| Mouse Expression Set 430 | 1417351_a_at | - | - |
| Mouse Expression Set 430 | 1417351_PM_a_at | - | - |
| Murine Genome U74v2 Set | 101506_PM_at | - | - |
| Murine Genome U74v2 Set | 101506_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417351_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417352_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417351_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417353_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417352_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417353_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417353_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417352_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417352_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417353_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1050735 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6110131 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050735 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110131 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110131 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773880 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732827 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773880 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2732827 | - | - |
| mouse-6 v1.1 (Illumina) | scl000146.1_5-S | - | - |
| mouse-6 v1.1 (Illumina) | scl32571.8.1_129-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050735 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110131 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000146.1_5-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32571.8.1_129-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110131 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050735 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2773880 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2732827 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2773880 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732827 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000146.1_5-S | - | - |

Snrpa1
Genomic Map