Assay Details
| Gene Symbol: Scn5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253860.1 | NP_001240789.1 | 9 - 10 | 1394 | 69 | |
| NM_021544.4 | NP_067519.2 | 10 - 11 | 1562 | 69 | ||
| GenBank mRNA | AJ271477.1 | - | 9 - 10 | 1390 | 69 | |
| AK147254.1 | - | 10 - 11 | 1562 | 69 | ||
| AK147517.1 | - | 10 - 11 | 1562 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scn5a
Genomic Map