Assay Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022018.3 | NP_071301.2 | 6 - 7 | 869 | 71 | |
| GenBank mRNA | AB049355.1 | - | 6 - 7 | 709 | 71 | |
| AK029847.1 | - | 2 - 3 | 181 | 71 | ||
| AK136597.1 | - | 6 - 7 | 779 | 71 | ||
| AK170193.1 | - | 6 - 7 | 792 | 71 | ||
| BC021332.1 | - | 6 - 7 | 757 | 71 | ||
| BC038006.1 | - | 6 - 7 | 725 | 71 | ||
| BC058234.1 | - | 4 - 5 | 469 | 6306849 | 71 | |
| BC099690.1 | - | 6 - 7 | 777 | 71 | ||
| BC137843.1 | - | 2 - 3 | 246 | 8860862 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam129a
Genomic Map