Assay Details
| Gene Symbol: Scamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022813.3 | NP_073724.1 | 3 - 4 | 331 | 90 | |
| GenBank mRNA | AF295402.1 | - | 3 - 4 | 335 | 90 | |
| AK003080.1 | - | 3 - 4 | 332 | 91 | ||
| AK161002.1 | - | 3 - 4 | 331 | 90 | ||
| AK171766.1 | - | 3 - 4 | 322 | 90 | ||
| BC014751.1 | - | 3 - 4 | 308 | 4235556 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scamp2
Genomic Map