Assay Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023799.3 | NP_076288.1 | 2 - 3 | 417 | 116 | |
| GenBank mRNA | AF132214.1 | - | 2 - 3 | 262 | 116 | |
| AK088774.1 | - | 2 - 3 | 680 | 116 | ||
| AK129188.1 | - | 2 - 3 | 646 | 116 | ||
| AK145227.1 | - | 2 - 3 | 667 | 116 | ||
| BC020093.1 | - | 2 - 3 | 354 | 116 | ||
| BC041109.1 | - | 2 - 3 | 747 | 116 | ||
| BC054821.1 | - | 2 - 3 | 417 | 6515250 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgea5
Genomic Map