Assay Details
| Gene Symbol: Trim39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024468.2 | NP_077788.2 | 5 - 6 | 922 | 75 | |
| NM_178281.1 | NP_840065.1 | 5 - 6 | 907 | 75 | ||
| GenBank mRNA | AB046382.1 | - | 5 - 6 | 922 | 75 | |
| AK145861.1 | - | 5 - 6 | 858 | 75 | ||
| AK166148.1 | - | 5 - 6 | 787 | 75 | ||
| AK220298.1 | - | 5 - 6 | 843 | 75 | ||
| BC031540.1 | - | 4 - 5 | 964 | 3666785 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim39
Genomic Map