Assay Details
| Gene Symbol: Ppil4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026141.3 | NP_080417.2 | 10 - 11 | 1040 | 93 | |
| GenBank mRNA | AK011443.1 | - | 10 - 11 | 1012 | 93 | |
| AK014406.2 | - | 1013 | 93 | |||
| AK017370.1 | - | 10 - 11 | 1040 | 93 | ||
| AK167258.1 | - | 10 - 11 | 1012 | 93 | ||
| BC004652.1 | - | 2 - 3 | 184 | 93 | ||
| BC079912.1 | - | 10 - 11 | 1002 | 30661438 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppil4
Genomic Map