Assay Details
| Gene Symbol: Mrpl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026759.3 | NP_081035.1 | 1 - 2 | 58 | 86 | |
| GenBank mRNA | AK003283.2 | - | 1 - 2 | 59 | 86 | |
| AK160744.1 | - | 1 - 2 | 55 | 86 | ||
| BC027542.1 | - | 1 - 2 | 42 | 86 | ||
| BC048498.1 | - | 1 - 2 | 61 | 86 | ||
| BC060968.1 | - | 1 - 2 | 64 | 6741102 | 86 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK003283.2,BC027542.1,BC060968.1,NM_026759.3 transcripts.
Set Membership: |
|

Mrpl13
Genomic Map