Assay Details
| Gene Symbol: Serpinb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027548.3 | NP_081824.1 | 4 - 5 | 402 | 104 | |
| GenBank mRNA | AF105328.1 | - | 3 - 4 | 340 | 104 | |
| AK014524.1 | - | 4 - 5 | 405 | 104 | ||
| BC064004.1 | - | 4 - 5 | 395 | 30288309 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb7
Genomic Map