Assay Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029391.2 | NP_083667.1 | 5 - 6 | 572 | 97 | |
| GenBank mRNA | AB232592.1 | - | 5 - 6 | 431 | 97 | |
| AF408432.1 | - | 5 - 6 | 544 | 97 | ||
| AK005314.1 | - | 5 - 6 | 572 | 97 | ||
| AK140513.1 | - | 4 - 5 | 769 | 97 | ||
| AK170552.1 | - | 5 - 6 | 586 | 97 | ||
| BC007147.1 | - | 5 - 6 | 516 | 3600193 | 97 | |
| BC096654.1 | - | 5 - 6 | 555 | 5056766 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab4b
Genomic Map