Assay Details
| Gene Symbol: Upf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122829.1 | NP_001116301.1 | 4 - 5 | 895 | 83 | |
| NM_030680.2 | NP_109605.2 | 4 - 5 | 895 | 83 | ||
| GenBank mRNA | AF182947.1 | - | 5 - 6 | 884 | 83 | |
| AF322655.1 | - | 4 - 5 | 615 | 83 | ||
| AK148196.1 | - | 5 - 6 | 872 | 83 | ||
| AK200037.1 | - | 1 - 2 | 61 | 83 | ||
| BC052149.1 | - | 4 - 5 | 833 | 6332288 | 83 | |
| BC056442.1 | - | 4 - 5 | 895 | 5702493 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upf1
Genomic Map