Assay Details
| Gene Symbol: Ntng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030699.2 | NP_109624.1 | 4 - 5 | 2155 | 74 | |
| GenBank mRNA | AB038667.1 | - | 4 - 5 | 1932 | 74 | |
| AF475069.1 | - | 3 - 4 | 1060 | 74 | ||
| AF475075.1 | - | 3 - 4 | 1060 | 74 | ||
| AF475076.1 | - | 3 - 4 | 1060 | 74 | ||
| AK048796.1 | - | 3 - 4 | 1021 | 74 | ||
More Information
Set Membership: |
|

Ntng1
Genomic Map