Assay Details
| Gene Symbol: Ndst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293682.1 | NP_001280611.1 | 5 - 6 | 763 | 86 | |
| NM_031186.3 | NP_112463.2 | 6 - 7 | 1876 | 86 | ||
| NR_121616.1 | 7 - 8 | 2024 | 86 | |||
| GenBank mRNA | AF221095.1 | - | 6 - 7 | 1875 | 86 | |
| AK014990.1 | - | 4 - 5 | 916 | 86 | ||
| AK015768.1 | - | 5 - 6 | 763 | 86 | ||
| AK141945.1 | - | 7 - 8 | 2025 | 86 | ||
| BC079622.1 | - | 6 - 7 | 1791 | 30535990 | 86 | |
| BC112404.1 | - | 6 - 7 | 1788 | 40060424 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndst3
Genomic Map