Assay Details
| Gene Symbol: Tyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282014.1 | NP_001268943.1 | 4 - 5 | 1192 | 63 | |
| NM_001282015.1 | NP_001268944.1 | 4 - 5 | 1186 | 63 | ||
| NM_031202.3 | NP_112479.1 | 4 - 5 | 1207 | 63 | ||
| GenBank mRNA | AK148332.1 | - | 4 - 5 | 1192 | 63 | |
| AK148370.1 | - | 4 - 5 | 1098 | 63 | ||
| AK148441.1 | - | 4 - 5 | 1098 | 63 | ||
| AK180622.1 | - | 1 - 2 | 109 | 63 | ||
| AK190391.1 | - | 1 - 2 | 109 | 63 | ||
| AK190783.1 | - | 1 - 2 | 109 | 63 | ||
| AK191129.1 | - | 1 - 2 | 109 | 63 | ||
| AK193049.1 | - | 1 - 2 | 109 | 63 | ||
| AK194119.1 | - | 109 | 63 | |||
| AK194181.1 | - | 1 - 2 | 109 | 63 | ||
| AK194480.1 | - | 1 - 2 | 109 | 63 | ||
| AK194755.1 | - | 1 - 2 | 83 | 63 | ||
| AK196212.1 | - | 1 - 2 | 109 | 63 | ||
| AK196465.1 | - | 1 - 2 | 109 | 63 | ||
| AK197240.1 | - | 1 - 2 | 109 | 63 | ||
| AK197400.1 | - | 1 - 2 | 109 | 63 | ||
| AK197957.1 | - | 1 - 2 | 109 | 63 | ||
| AK199510.1 | - | 1 - 2 | 109 | 63 | ||
| AK199962.1 | - | 1 - 2 | 109 | 63 | ||
| AK200845.1 | - | 1 - 2 | 109 | 63 | ||
| AK201045.1 | - | 1 - 2 | 109 | 63 | ||
| AK201297.1 | - | 1 - 2 | 109 | 63 | ||
| AK203202.1 | - | 1 - 2 | 109 | 63 | ||
| AK203316.1 | - | 109 | 63 | |||
| AK204783.1 | - | 1 - 2 | 109 | 63 | ||
| AK211654.1 | - | 1 - 2 | 109 | 63 | ||
| AK214877.1 | - | 1 - 2 | 109 | 63 | ||
| BC076598.1 | - | 4 - 5 | 1190 | 30613975 | 63 | |
| X03687.1 | - | 4 - 5 | 1092 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tyrp1
Genomic Map