Assay Details
| Gene Symbol: Gigyf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031408.2 | NP_113596.2 | 16 - 17 | 2029 | 83 | |
| GenBank mRNA | AK220289.1 | - | 9 - 10 | 1038 | 83 | |
| AY176042.1 | - | 16 - 17 | 2045 | 83 | ||
| BC052875.1 | - | 10 - 11 | 1133 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gigyf1
Genomic Map