Assay Details
| Gene Symbol: Supt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033618.3 | NP_291096.2 | 20 - 21 | 2505 | 75 | |
| GenBank mRNA | AF323667.1 | - | 20 - 21 | 2467 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt16
Genomic Map