Assay Details
| Gene Symbol: Recql4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058214.3 | NP_478121.2 | 7 - 8 | 1432 | 77 | |
| GenBank mRNA | AB039882.1 | - | 7 - 8 | 1305 | 77 | |
| AB175741.1 | - | 7 - 8 | 1349 | 77 | ||
| BC064680.1 | - | 7 - 8 | 1406 | 77 | ||
| BC131656.1 | - | 7 - 8 | 1345 | 77 | ||
| BC131657.1 | - | 6 - 7 | 1532 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Recql4
Genomic Map