Assay Details
| Gene Symbol: Recql4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058214.3 | NP_478121.2 | 11 - 12 | 1839 | 92 | |
| GenBank mRNA | AB039882.1 | - | 11 - 12 | 1712 | 92 | |
| AB175741.1 | - | 11 - 12 | 1756 | 92 | ||
| AK090260.1 | - | 12 - 13 | 1443 | 92 | ||
| BC064680.1 | - | 11 - 12 | 1813 | 92 | ||
| BC131656.1 | - | 11 - 12 | 1752 | 92 | ||
| BC131657.1 | - | 10 - 11 | 1939 | 92 | ||
More Information
Set Membership: |
|

Recql4
Genomic Map