Assay Details
| Gene Symbol: Recql4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058214.3 | NP_478121.2 | 15 - 16 | 2618 | 62 | |
| GenBank mRNA | AB175741.1 | - | 15 - 16 | 2535 | 62 | |
| AK090260.1 | - | 16 - 17 | 2223 | 62 | ||
| BC064680.1 | - | 15 - 16 | 2592 | 62 | ||
| BC131656.1 | - | 15 - 16 | 2531 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Recql4
Genomic Map