Assay Details
| Gene Symbol: Sardh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138665.2 | NP_619606.1 | 3 - 4 | 1184 | 72 | |
| GenBank mRNA | AK140451.1 | - | 2 - 3 | 460 | 72 | |
| AK159236.1 | - | 3 - 4 | 1148 | 72 | ||
| AK159681.1 | - | 3 - 4 | 1148 | 72 | ||
| AK159704.1 | - | 3 - 4 | 1148 | 72 | ||
| AK163515.1 | - | 3 - 4 | 713 | 72 | ||
| AK163660.1 | - | 2 - 3 | 460 | 72 | ||
| BC003456.1 | - | 3 - 4 | 693 | 2648344 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sardh
Genomic Map