Assay Details
| Gene Symbol: Shkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138676.2 | NP_619617.2 | 3 - 4 | 235 | 95 | |
| GenBank mRNA | AF246218.1 | - | 3 - 4 | 241 | 95 | |
| AK047434.1 | - | 1 - 2 | 308 | 95 | ||
| AK154779.1 | - | 3 - 4 | 224 | 95 | ||
| AK160641.1 | - | 2 - 3 | 307 | 95 | ||
| AK166328.1 | - | 2 - 3 | 180 | 95 | ||
| BC061477.1 | - | 3 - 4 | 188 | 6336438 | 95 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK160641.1 transcripts.
Set Membership: |
|

Shkbp1
Genomic Map