Assay Details
| Gene Symbol: Shkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138676.2 | NP_619617.2 | 8 - 9 | 705 | 85 | |
| GenBank mRNA | AF246218.1 | - | 8 - 9 | 711 | 85 | |
| AK047434.1 | - | 6 - 7 | 836 | 85 | ||
| AK154779.1 | - | 8 - 9 | 694 | 85 | ||
| AK160641.1 | - | 7 - 8 | 777 | 85 | ||
| AK190932.1 | - | 2 - 3 | 230 | 85 | ||
| BC061477.1 | - | 8 - 9 | 658 | 6336438 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shkbp1
Genomic Map