Assay Details
| Gene Symbol: Shkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138676.2 | NP_619617.2 | 14 - 15 | 1546 | 82 | |
| GenBank mRNA | AF246218.1 | - | 14 - 15 | 1552 | 82 | |
| AK047434.1 | - | 12 - 13 | 1677 | 82 | ||
| AK047826.1 | - | 2 - 3 | 345 | 82 | ||
| AK154779.1 | - | 14 - 15 | 1534 | 82 | ||
| AK160641.1 | - | 13 - 14 | 1618 | 82 | ||
| AK182187.1 | - | 1 - 2 | 77 | 82 | ||
| AK186318.1 | - | 1 - 2 | 117 | 82 | ||
| AK187576.1 | - | 1 - 2 | 77 | 82 | ||
| BC061477.1 | - | 14 - 15 | 1499 | 6336438 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shkbp1
Genomic Map