Assay Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138758.1 | NP_620097.1 | 7 - 8 | 1260 | 73 | |
| GenBank mRNA | AK144627.1 | - | 7 - 8 | 1256 | 73 | |
| AK161724.1 | - | 7 - 8 | 1227 | 73 | ||
| AY033513.1 | - | 6 - 7 | 1098 | 73 | ||
| BC010495.1 | - | 7 - 8 | 1260 | 4195680 | 73 | |
| BC115365.1 | - | 7 - 8 | 1192 | 40059106 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carnitine biosynthetic process
negative regulation of oxidoreductase activity
oxidation-reduction process
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
L-ascorbic acid binding
metal ion binding
trimethyllysine dioxygenase activity
dioxygenase activity
carnitine biosynthetic process
negative regulation of oxidoreductase activity
oxidation-reduction process
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
L-ascorbic acid binding
metal ion binding
trimethyllysine dioxygenase activity
dioxygenase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | BC010495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | BC115365.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | AK144627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | AK161724.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | NM_138758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | BC010495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | BC115365.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | AK144627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | AK161724.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509452 | NM_138758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | BC010495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | BC115365.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | AK144627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | AK161724.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | NM_138758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | BC010495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | BC115365.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | AK144627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | AK161724.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509452 | NM_138758.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | BC010495.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | BC115365.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | AK144627.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | AK161724.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | NM_138758.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | BC010495.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | BC115365.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | AK144627.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | AK161724.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509452 | NM_138758.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577766 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577743 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577758 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577753 | - | - |
| Whole Mouse Genome G4122A | A_51_P509452 | BC010495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | BC115365.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | AK144627.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | AK161724.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | NM_138758.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | BC010495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | BC115365.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | AK144627.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | AK161724.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509452 | NM_138758.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112828 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09577761 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09577743 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10608712 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420727_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420725_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420726_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608712 | - | - |
| Mouse Genome 430 2.0 Array | 1452500_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420727_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1446546_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446546_at | - | - |
| Mouse Genome 430 2.0 Array | 1420727_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420726_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452500_at | - | - |
| Mouse Genome 430 2.0 Array | 1420725_at | - | - |
| Mouse Genome 430 2.0 Array | 1420726_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1420725_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSRX_GL456233_random00000012.mm.1 | NM_138758.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSRX_GL456233_random00000012.mm.1 | TCX_GL456233_random00000002.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSRX_GL456233_random00000013.mm.1 | NM_138758.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSRX_GL456233_random00000013.mm.1 | TCX_GL456233_random00000002.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420727_PM_a_at | - | - |
| Mouse Expression Set 430 | 1420727_a_at | - | - |
| Mouse Expression Set 430 | 1420726_x_at | - | - |
| Mouse Expression Set 430 | 1420725_at | - | - |
| Mouse Expression Set 430 | 1420725_PM_at | - | - |
| Mouse Expression Set 430 | 1420726_PM_x_at | - | - |
| Mouse Expression Set 430 | 1446546_PM_at | - | - |
| Mouse Expression Set 430 | 1446546_at | - | - |
| Murine Genome U74v2 Set | 138802_at | - | - |
| Murine Genome U74v2 Set | 138802_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420725_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420726_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420727_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420725_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420726_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420727_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420726_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420727_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420726_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420725_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420727_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420725_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5420136 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2513145 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2513145 | - | - |
| mouse-6 v1.1 (Illumina) | scl00192289.1_110-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00192289.1_110-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2513145 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2513145 | - | - |

Tmlhe
Genomic Map