Assay Details
| Gene Symbol: Vwa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147776.4 | NP_680085.3 | 2 - 3 | 692 | 91 | |
| GenBank mRNA | AK030019.1 | - | 2 - 3 | 646 | 91 | |
| AK077240.1 | - | 2 - 3 | 663 | 91 | ||
| AY030094.1 | - | 2 - 3 | 662 | 91 | ||
| BC026919.1 | - | 2 - 3 | 651 | 3982749 | 91 | |
| BC036166.1 | - | 2 - 3 | 657 | 4985003 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vwa1
Genomic Map