Assay Details
| Gene Symbol: Sftpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282071.2 | NP_001269000.1 | 2 - 3 | 205 | 73 | |
| NM_147779.2 | NP_680088.1 | 2 - 3 | 205 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sftpb
Genomic Map