Assay Details
| Gene Symbol: Sftpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282071.2 | NP_001269000.1 | 5 - 6 | 569 | 110 | |
| NM_147779.2 | NP_680088.1 | 5 - 6 | 569 | 110 | ||
| GenBank mRNA | AI430444.1 | - | 2 - 3 | 222 | 110 | |
| AI894333.1 | - | 2 - 3 | 297 | 110 | ||
More Information
Set Membership: |
|

Sftpb
Genomic Map