Assay Details
| Gene Symbol: Sftpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147779.2 | NP_680088.1 | 10 - 11 | 1164 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sftpb| Gene Symbol: Sftpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147779.2 | NP_680088.1 | 10 - 11 | 1164 | 65 | |
Set Membership: |
|