Assay Details
| Gene Symbol: Rps6ka3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148945.2 | NP_683747.1 | 2 - 3 | 466 | 95 | |
| GenBank mRNA | AK079102.1 | - | 559 | 95 | ||
| AK154778.1 | - | 2 - 3 | 379 | 95 | ||
| AY083469.1 | - | 2 - 3 | 128 | 95 | ||
| BC150156.1 | - | 2 - 3 | 131 | 95 | ||
| BC150478.1 | - | 2 - 3 | 131 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka3
Genomic Map