Assay Details
| Gene Symbol: Rps6ka3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148945.2 | NP_683747.1 | 12 - 13 | 1330 | 99 | |
| GenBank mRNA | AK154778.1 | - | 12 - 13 | 1243 | 99 | |
| AY083469.1 | - | 12 - 13 | 992 | 99 | ||
| BC150156.1 | - | 12 - 13 | 995 | 99 | ||
| BC150478.1 | - | 12 - 13 | 995 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka3
Genomic Map