Assay Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029838.2 | NP_001025009.1 | 6 - 7 | 678 | 76 | |
| NM_148950.3 | NP_683752.2 | 5 - 6 | 573 | 76 | ||
| GenBank mRNA | AB086817.1 | - | 3 - 4 | 399 | 76 | |
| AF487460.1 | - | 4 - 5 | 479 | 76 | ||
| AK082952.1 | - | 6 - 7 | 623 | 76 | ||
| AK084178.1 | - | 6 - 7 | 621 | 76 | ||
| BC050865.1 | - | 5 - 6 | 507 | 6332968 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pknox2
Genomic Map