Assay Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029838.2 | NP_001025009.1 | 9 - 10 | 1089 | 79 | |
| NM_148950.3 | NP_683752.2 | 8 - 9 | 984 | 79 | ||
| GenBank mRNA | AB086817.1 | - | 6 - 7 | 810 | 79 | |
| AF487460.1 | - | 7 - 8 | 890 | 79 | ||
| AK082952.1 | - | 9 - 10 | 1034 | 79 | ||
| BC050865.1 | - | 8 - 9 | 918 | 6332968 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pknox2
Genomic Map