Assay Details
| Gene Symbol: Fscn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007984.2 | NP_032010.2 | 3 - 4 | 1205 | 94 | |
| GenBank mRNA | AK083935.1 | - | 3 - 4 | 1206 | 94 | |
| AK167670.1 | - | 3 - 4 | 1205 | 94 | ||
| BC010338.1 | - | 3 - 4 | 1191 | 94 | ||
| BC029281.1 | - | 3 - 4 | 1193 | 94 | ||
| BC037137.1 | - | 3 - 4 | 1202 | 94 | ||
| BC052408.1 | - | 3 - 4 | 1194 | 5716179 | 94 | |
| L33726.1 | - | 3 - 4 | 1189 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fscn1
Genomic Map