Assay Details
| Gene Symbol: St8sia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009181.2 | NP_033207.2 | 2 - 3 | 343 | 124 | |
| GenBank mRNA | BC096546.1 | - | 2 - 3 | 335 | 6833487 | 124 |
| X83562.1 | - | 2 - 3 | 322 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

St8sia2
Genomic Map