Assay Details
| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009417.2 | NP_033443.1 | 4 - 5 | 407 | 99 | |
| GenBank mRNA | AK047843.1 | - | 4 - 5 | 386 | 99 | |
| BC099421.1 | - | 3 - 4 | 339 | 99 | ||
| X60703.1 | - | 4 - 5 | 407 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpo
Genomic Map