Assay Details
| Gene Symbol: Zfp57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D21850.1 | - | 1 - 2 | 283 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D21850.1 transcripts.
Set Membership: |
|

Zfp57
Genomic Map