| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200022061.mm.1 |
NM_019690.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022061.mm.1 |
NM_022000.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022061.mm.1 |
TC0200002700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200022058.mm.1 |
NM_019690.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022058.mm.1 |
NM_022000.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022058.mm.1 |
TC0200002700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200022059.mm.1 |
NM_019690.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022059.mm.1 |
NM_022000.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022059.mm.1 |
TC0200002700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200022060.mm.1 |
NM_019690.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022060.mm.1 |
NM_022000.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022060.mm.1 |
TC0200002700.mm.1 |
MTA TC |