Assay Details
| Gene Symbol: Gsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206367.1 | NP_001193296.1 | 5 - 6 | 721 | 63 | |
| NM_001206368.1 | NP_001193297.1 | 4 - 5 | 637 | 63 | ||
| NM_001206369.1 | NP_001193298.1 | 4 - 5 | 682 | 63 | ||
| NM_146120.4 | NP_666232.2 | 4 - 5 | 744 | 63 | ||
| GenBank mRNA | AK076156.1 | - | 4 - 5 | 684 | 63 | |
| AK089934.1 | - | 5 - 6 | 676 | 63 | ||
| AK135632.1 | - | 3 - 4 | 457 | 63 | ||
| AK143664.1 | - | 4 - 5 | 711 | 63 | ||
| AK146969.1 | - | 5 - 6 | 638 | 63 | ||
| AK148957.1 | - | 5 - 6 | 701 | 63 | ||
| AK151684.1 | - | 5 - 6 | 663 | 63 | ||
| AK153029.1 | - | 5 - 6 | 641 | 63 | ||
| AK166831.1 | - | 4 - 5 | 657 | 63 | ||
| AK166932.1 | - | 5 - 6 | 635 | 63 | ||
| AK167918.1 | - | 5 - 6 | 651 | 63 | ||
| AK168461.1 | - | 5 - 6 | 654 | 63 | ||
| AK168532.1 | - | 5 - 6 | 655 | 63 | ||
| AK168707.1 | - | 4 - 5 | 590 | 63 | ||
| AK168725.1 | - | 5 - 6 | 638 | 63 | ||
| AK168829.1 | - | 4 - 5 | 689 | 63 | ||
| AK169041.1 | - | 5 - 6 | 641 | 63 | ||
| AK169670.1 | - | 5 - 6 | 666 | 63 | ||
| BC023143.1 | - | 4 - 5 | 682 | 3711602 | 63 | |
| BC060377.1 | - | 5 - 6 | 669 | 4983664 | 63 | |
| J04953.1 | - | 3 - 4 | 531 | 63 | ||
More Information
Set Membership: |
|

Gsn
Genomic Map