| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400014064.mm.1 |
NM_001190978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014064.mm.1 |
NM_001190978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014064.mm.1 |
NM_010623.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014064.mm.1 |
NM_010623.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014064.mm.1 |
TC0400001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014060.mm.1 |
NM_001190978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014060.mm.1 |
NM_001190978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014060.mm.1 |
NM_010623.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014060.mm.1 |
NM_010623.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014060.mm.1 |
TC0400001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014063.mm.1 |
NM_001190978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014063.mm.1 |
NM_001190978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014063.mm.1 |
NM_010623.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014063.mm.1 |
NM_010623.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014063.mm.1 |
TC0400001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014061.mm.1 |
NM_001190978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014061.mm.1 |
NM_001190978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014061.mm.1 |
NM_010623.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014061.mm.1 |
NM_010623.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014061.mm.1 |
TC0400001721.mm.1 |
MTA TC |