Assay Details
| Gene Symbol: Kif17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010623.4 | NP_034753.1 | 6 - 7 | 1481 | 78 | |
| GenBank mRNA | AB008867.1 | - | 6 - 7 | 1479 | 78 | |
| AK046890.1 | - | 6 - 7 | 1234 | 78 | ||
More Information
Set Membership: |
|

Kif17
Genomic Map