Assay Details
| Gene Symbol: Ncr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010746.3 | NP_034876.2 | 1 - 2 | 56 | 79 | |
| GenBank mRNA | AJ223765.1 | - | 1 - 2 | 45 | 79 | |
| AK079401.1 | - | 1 - 2 | 57 | 79 | ||
| BC115364.2 | - | 1 - 2 | 57 | 40045898 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ223765.1, AK079401.1, BC115364.2, NM_010746.3 transcripts.
Set Membership: |
|

Ncr1
Genomic Map