Assay Details
| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010218.1 | - | 1 - 2 | 233 | 72 | |
| AK038631.1 | - | 1 - 2 | 281 | 72 | ||
| AK156731.1 | - | 1 - 2 | 207 | 72 | ||
| AY176058.1 | - | 1 - 2 | 240 | 72 | ||
| AY176059.1 | - | 1 - 2 | 271 | 72 | ||
| BC021938.1 | - | 1 - 2 | 238 | 5136809 | 72 | |
| U19799.1 | - | 1 - 2 | 252 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkbib
Genomic Map