Assay Details
| Gene Symbol: Rhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011270.3 | NP_035400.3 | 2 - 3 | 402 | 68 | |
| GenBank mRNA | AB015189.1 | - | 2 - 3 | 337 | 68 | |
| AF012430.1 | - | 2 - 3 | 317 | 68 | ||
| AF047827.1 | - | 2 - 3 | 337 | 68 | ||
| AF057524.1 | - | 2 - 3 | 354 | 68 | ||
| AF069311.1 | - | 2 - 3 | 356 | 68 | ||
| AK079335.1 | - | 2 - 3 | 346 | 68 | ||
| AK089642.1 | - | 2 - 3 | 355 | 68 | ||
| AK157850.1 | - | 2 - 3 | 329 | 68 | ||
| AK171641.1 | - | 2 - 3 | 343 | 68 | ||
| BC145805.1 | - | 2 - 3 | 433 | 40131108 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhd
Genomic Map