Assay Details
| Gene Symbol: Vrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029843.1 | NP_001025014.1 | 9 - 10 | 928 | 140 | |
| NM_001029844.1 | NP_001025015.1 | 9 - 10 | 928 | 140 | ||
| NM_011705.3 | NP_035835.1 | 9 - 10 | 928 | 140 | ||
| GenBank mRNA | AF080252.1 | - | 8 - 9 | 841 | 140 | |
| AF080253.1 | - | 9 - 10 | 882 | 140 | ||
| AK032344.1 | - | 9 - 10 | 899 | 140 | ||
| AK088230.1 | - | 9 - 10 | 852 | 140 | ||
| AK088674.1 | - | 9 - 10 | 911 | 140 | ||
| AK136352.1 | - | 9 - 10 | 902 | 140 | ||
| AK166676.1 | - | 9 - 10 | 896 | 140 | ||
| BC016676.1 | - | 9 - 10 | 963 | 3602292 | 140 | |
| BC051072.1 | - | 9 - 10 | 908 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vrk1
Genomic Map